CAS RN 14564-74-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Dodecylbenzenesulfonic Acid Potassium Salt, TRC
CAS: 14564-74-0 Molecular Formula: C18H29KO3S Molecular Weight (g/mol): 364.58 Synonym: p-Dodecylbenzenesulfonic Acid Potassium Salt SMILES: [K+].CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)[O-]