accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3OHO

CAS RN 14573-97-8

IUPAC Name: {2-[3-(dimethylamino)propoxy]phenyl}methanol
Synonyms: {2-[3-(dimethylamino)propoxy]phenyl}methanol
Molecular Weight (g/mol): 209.29
Molecular Formula: C12H19NO2
InChi Key: KEQGOXOOPXJVFM-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results