accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 145731-49-3

IUPAC Name: benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Synonyms: benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate
Molecular Weight (g/mol): 504.63
Molecular Formula: C26H40N4O6
InChi Key: TZVQRMYLYQNBOA-UHFFFAOYNA-N
SMILES: CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results