accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2CBr

CAS RN 1458-98-6

IUPAC Name: 3-bromo-2-methylprop-1-ene
Synonyms: 3-bromo-2-methylprop-1-ene
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: USEGQJLHQSTGHW-UHFFFAOYSA-N
SMILES: CC(=C)CBr
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)