accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 1459-10-5

IUPAC Name: tetradecylbenzene
Synonyms: 1-phenyltetradecane tetradecane,phenyl
Molecular Weight (g/mol): 274.49
Molecular Formula: C20H34
InChi Key: JZALLXAUNPOCEU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)