accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CH3CCH3O

CAS RN 14593-46-5

IUPAC Name: sodium 2,2-dimethylpropan-1-olate
Synonyms: sodium 2,2-dimethylpropan-1-olate
Molecular Weight (g/mol): 110.13
Molecular Formula: C5H11NaO
InChi Key: YQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES: [Na+].CC(C)(C)C[O-]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)