accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 146-68-9

IUPAC Name: 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: int (chemical)
Molecular Weight (g/mol): 505.7
Molecular Formula: C19H13ClIN5O2
InChi Key: JORABGDXCIBAFL-UHFFFAOYSA-M
SMILES: [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)