accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClSnHH3CH3CH3C

CAS RN 1461-22-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 327.52
Molecular Formula: C12H29ClSn
InChi Key: UEKQSLXYFDHEJR-UHFFFAOYSA-N
SMILES: Cl.CCCC[SnH](CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (7)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (7)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)