accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14610-37-8

H3CNH2CH3CH3H3C

CAS RN 14610-37-8

IUPAC Name: tert-butyl(methyl)azanium
Synonyms: tert-butyl(methyl)azanium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: ZQGJEUVBUVKZKS-UHFFFAOYSA-O
SMILES: C[NH2+]C(C)(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)