accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOFFF

CAS RN 146137-78-2

IUPAC Name: 2-fluoro-5-(trifluoromethyl)benzaldehyde
Synonyms: 2-fluoro-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: IDLNLGMUINCSGS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1C=O)C(F)(F)F
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results