accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14647-23-5

IUPAC Name: nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
Synonyms: nickel(2+) [2-(diphenylphosphaniumyl)ethyl]diphenylphosphanium dichloride
Molecular Weight (g/mol): 530.03
Molecular Formula: C26H26Cl2NiP2
InChi Key: XXECWTBMGGXMKP-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[Ni++].C(C[PH+](C1=CC=CC=C1)C1=CC=CC=C1)[PH+](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)