accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14653-77-1

HHClClH2NNHSH

CAS RN 14653-77-1

IUPAC Name: dihydrogen 2-[(3-aminopropyl)amino]ethane-1-thiol dichloride
Synonyms: dihydrogen 2-[(3-aminopropyl)amino]ethanethiol dichloride
Molecular Weight (g/mol): 207.16
Molecular Formula: C5H16Cl2N2S
InChi Key: XDRLRDHLCIFZIW-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCCNCCS
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results