accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOO(E)

CAS RN 1466-88-2

IUPAC Name: (2E)-3-(2-nitrophenyl)prop-2-enal
Synonyms: 2-nitrocinnamaldehyde
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: VMSMELHEXDVEDE-HWKANZROSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1\C=C\C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results