accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaONOONNONO(E)

CAS RN 14663-23-1

IUPAC Name: sodium 3-[(E)-{[5-(4-nitrophenyl)furan-2-yl]methylidene}amino]-2,5-dioxoimidazolidin-1-ide
Synonyms: sodium 3-[(E)-{[5-(4-nitrophenyl)furan-2-yl]methylidene}amino]-2,5-dioxoimidazolidin-1-ide
Molecular Weight (g/mol): 336.24
Molecular Formula: C14H9N4NaO5
InChi Key: KSRLIXGNPXAZHD-HAZZGOGXSA-M
SMILES: [Na+].[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results