CAS RN 146724-94-9
IUPAC Name:
N-[bis(2-azaniumylethyl)amino]-N-nitrosohydroxylamine
Synonyms:
N-[bis(2-ammonioethyl)amino]-N-nitrosohydroxylamine
Molecular Weight (g/mol):
165.2
Molecular Formula:
C4H15N5O2
InChi Key:
HMRRJTFDJAVRMR-UHFFFAOYSA-P
SMILES:
[NH3+]CCN(CC[NH3+])N(O)N=O
1
–
1
of
1
results
1
Promotions Available
DETA NONOate, 95%
DETA NONOate, 95%
CAS: 146724-94-9 Molecular Formula: C4H13N5O2 Molecular Weight (g/mol): 163.2 MDL Number: MFCD00278808 InChI Key: HMRRJTFDJAVRMR-UHFFFAOYSA-N Synonym: deta nonoate,noc-18,deta-nonoate,diethylenetriamine nonoate,2,2'-hydroxynitrosohydrazino bis-ethanamine,aeed/no,1,1-bis 2-aminoethyl-2-hydroxy-3-oxotriazane,3,3-bis aminoethyl-1-hydroxy-2-oxo-1-triazene,detanonoate,1-hydroxy-2-oxo-3,3-bis 3-aminoethyl-1-triazene PubChem CID: 1606 ChEBI: CHEBI:50154 IUPAC Name: N-[bis(2-aminoethyl)amino]-N-hydroxynitrous amide SMILES: C(CN(CCN)N(N=O)O)N