accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1468-95-7

IUPAC Name: (anthracen-9-yl)methanol
Synonyms: 9-anthracenemethanol
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: JCJNNHDZTLRSGN-UHFFFAOYSA-N
SMILES: OCC1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)