accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1469-48-3

IUPAC Name: (3aR,7aS)-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione
Synonyms: (3aR,7aS)-3a,4,7,7a-tetrahydro-2H-isoindole-1,3-dione
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: CIFFBTOJCKSRJY-OLQVQODUSA-N
SMILES: O=C1NC(=O)[C@@H]2CC=CC[C@H]12
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results