accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OPOOOIn

CAS RN 14693-82-4

IUPAC Name: indium(3+) phosphate
Synonyms: indium(3+) phosphate
Molecular Weight (g/mol): 209.79
Molecular Formula: InO4P
InChi Key: UJXZVRRCKFUQKG-UHFFFAOYSA-K
SMILES: [In+3].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results