accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHOOHOHOHHH

CAS RN 147-71-7

IUPAC Name: (2S,3S)-2,3-dihydroxybutanedioic acid
Synonyms: D-tartaric acid tartaric acid, D(-)- D-(-)-tartaric acid linksweinsaeure (-)-(S,S)-tartaric acid (-)-D-tartaric acid (-)-weinsaeure D(-)-tartaric acid d(-)-tartaric acid (S,S)-(-)-tartaric acid
Molecular Weight (g/mol): 150.09
Molecular Formula: C4H6O6
InChi Key: FEWJPZIEWOKRBE-LWMBPPNESA-N
SMILES: O[C@@H]([C@H](O)C(O)=O)C(O)=O
Percent Purity 
  • (4)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)