accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOHN

CAS RN 147-85-3

IUPAC Name: pyrrolidine-2-carboxylic acid
Synonyms: proline prolin Hpro proline DL-form
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1
Quantity 
  • (1)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
Grade 
  • (1)
  • (4)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)