accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 147059-75-4

CH3SOHOOH2N(S)N(R)NNOHOOFFF

CAS RN 147059-75-4

IUPAC Name: 7-[(1R,5S)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid; methanesulfonic acid
Synonyms: methanesulfonic acid; trovafloxacin
Molecular Weight (g/mol): 512.46
Molecular Formula: C21H19F3N4O6S
InChi Key: DYNZICQDCVYXFW-GIPYJWDTSA-N
SMILES: CS(O)(=O)=O.NC1[C@@H]2CN(C[C@H]12)C1=C(F)C=C2C(=O)C(=CN(C3=CC=C(F)C=C3F)C2=N1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results