accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CPPCH3H3CCH3H3CH3CCH3

CAS RN 147253-69-8

IUPAC Name: 1-{2-[2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane
Synonyms: 1-[2-(2,5-diisopropylphospholan-1-yl)phenyl]-2,5-diisopropylphospholane
Molecular Weight (g/mol): 418.59
Molecular Formula: C26H44P2
InChi Key: RBVGOQHQBUPSGX-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C(C)C)P1C1=CC=CC=C1P1C(CCC1C(C)C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results