accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ZnSSNSSN

CAS RN 14726-36-4

IUPAC Name: zinc(2+) bis((dibenzylcarbamothioyl)sulfanide)
Synonyms: zinc(2+) bis((dibenzylcarbamothioyl)sulfanide)
Molecular Weight (g/mol): 610.19
Molecular Formula: C30H28N2S4Zn
InChi Key: AUMBZPPBWALQRO-UHFFFAOYSA-L
SMILES: [Zn++].[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1.[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)