accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBOHHN

CAS RN 147621-18-9

IUPAC Name: (1H-indol-6-yl)boronic acid
Synonyms: 1H-indol-6-ylboronic acid
Molecular Weight (g/mol): 160.97
Molecular Formula: C8H8BNO2
InChi Key: ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results