accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CONH2HO

CAS RN 1477-68-5

IUPAC Name: hydrogen 4-(2-aminoethyl)-2-methoxyphenol chloride
Synonyms: hydrogen methoxytyramine chloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: AWRIOTVUTPLWLF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC(CCN)=CC=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results