accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 147741-30-8

H3CNCH3H3CH3CH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OH2OOH

CAS RN 147741-30-8

IUPAC Name: tetrabutylazanium triacontahydrate hydroxide
Synonyms: tetrabutylammonium ion triacontahydrate hydroxide
Molecular Weight (g/mol): 799.93
Molecular Formula: C16H97NO31
InChi Key: DFGIRVKFXSRUOX-UHFFFAOYSA-M
SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results