accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14782-58-2

IUPAC Name: potassium tetrakis(1H-pyrazol-1-yloxy)boranuide
Synonyms: potassium tetrakis(pyrazol-1-yloxy)boranuide
Molecular Weight (g/mol): 382.19
Molecular Formula: C12H12BKN8O4
InChi Key: RKBGLZQVHBLUBM-UHFFFAOYSA-N
SMILES: [K+].O(N1C=CC=N1)[B-](ON1C=CC=N1)(ON1C=CC=N1)ON1C=CC=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)