accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH3COSn

CAS RN 14791-99-2

IUPAC Name: λ²-tin(2+) diethanolate
Synonyms: λ²-tin(2+) bis(ethoxide)
Molecular Weight (g/mol): 208.83
Molecular Formula: C4H10O2Sn
InChi Key: XCKWFNSALCEAPW-UHFFFAOYSA-N
SMILES: [Sn++].CC[O-].CC[O-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)