accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOO

CAS RN 14797-55-8

IUPAC Name: nitrate
Synonyms: nitrate nitrate(1-)
Molecular Weight (g/mol): 62.01
Molecular Formula: NO3
InChi Key: NHNBFGGVMKEFGY-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (5)
  • (1)
Concentration 
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (8)
  • (6)
  • (1)
  • (8)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)