accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14807-75-1

IUPAC Name: [(azaniumylmethanimidoyl)disulfanyl](imino)methanaminium dichloride
Synonyms: [(ammoniomethanimidoyl)disulfanyl](imino)methanaminium dichloride
Molecular Weight (g/mol): 223.13
Molecular Formula: C2H8Cl2N4S2
InChi Key: BFJQSCVWXZOXGK-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results