accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1481-41-0

CH3SiCH3ClFFF

CAS RN 1481-41-0

IUPAC Name: chlorodimethyl(3,3,3-trifluoropropyl)silane
Synonyms: chlorodimethyl(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol): 190.67
Molecular Formula: C5H10ClF3Si
InChi Key: KBAZUXSLKGQRJF-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCC(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results