accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClI

CAS RN 1483-72-3

IUPAC Name: diphenyliodanium chloride
Synonyms: diphenyliodanium chloride
Molecular Weight (g/mol): 316.57
Molecular Formula: C12H10ClI
InChi Key: RSJLWBUYLGJOBD-UHFFFAOYSA-M
SMILES: [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results