accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3C(E)

CAS RN 14850-23-8

IUPAC Name: (4E)-oct-4-ene
Synonyms: 4-octene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: IRUCBBFNLDIMIK-BQYQJAHWSA-N
SMILES: CCC\C=C\CCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results