accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 148546-97-8

CH3NNNOO

CAS RN 148546-97-8

IUPAC Name: 1-methyl-4-(3-nitrophenyl)piperazine
Synonyms: 1-methyl-4-(3-nitrophenyl)piperazine
Molecular Weight (g/mol): 221.26
Molecular Formula: C11H15N3O2
InChi Key: IIRKKCDXJIXWHI-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results