accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSiCH3H3C

CAS RN 14857-34-2

IUPAC Name: ethoxydimethylsilyl
Synonyms: ethoxydimethylsilyl
Molecular Weight (g/mol): 103.22
Molecular Formula: C4H11OSi
InChi Key: DRUOQOFQRYFQGB-UHFFFAOYSA-N
SMILES: CCO[Si](C)C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)