accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 148644-09-1

IUPAC Name: 3-methoxyazetidine hydrochloride
Synonyms: 3-methoxyazetidine hydrochloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: KSXGQRBTBLQJEF-UHFFFAOYSA-N
SMILES: Cl.COC1CNC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results