accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IrClCPPOPhPhPhPhPhPh

CAS RN 14871-41-1

IUPAC Name: iridium(1+) methanediidylidyneoxidanium bis(triphenylphosphane) hydrochloridyl
Synonyms: iridium(1+) methanediidylidyneoxidanium bis(triphenylphosphine) hydrochloridyl
Molecular Weight (g/mol): 780.26
Molecular Formula: C37H30ClIrOP2
InChi Key: NIQUQIBOURJEPY-UHFFFAOYSA-N
SMILES: [Cl].[Ir+].[C--]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results