accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNFFF

CAS RN 148853-42-3

IUPAC Name: 2-(trifluoromethyl)quinoxaline
Synonyms: 2-(trifluoromethyl)quinoxaline
Molecular Weight (g/mol): 198.15
Molecular Formula: C9H5F3N2
InChi Key: YIQVNKFWTSLVSA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC2=CC=CC=C2N=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results