accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 148890-63-5

IUPAC Name: 5-amino-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 5-amino-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: GXGYWLOHPYNWFE-UHFFFAOYSA-N
SMILES: NC1=C2NC(=O)COC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results