accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 148893-10-1

IUPAC Name: [bis(dimethylamino)methylidene]({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yl})oxidanium
Synonyms: HATU
Molecular Weight (g/mol): 235.27
Molecular Formula: C10H15N6O
InChi Key: WXIONIWNXBAHRU-UHFFFAOYSA-N
SMILES: CN(C)C(=[O+]N1N=NC2=CC=CN=C12)N(C)C
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results