accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 149-44-0

IUPAC Name: sodium hydroxymethanesulfinate
Synonyms: rongalite sodium oxymethylene sulfoxylate
Molecular Weight (g/mol): 118.08
Molecular Formula: CH3NaO3S
InChi Key: XWGJFPHUCFXLBL-UHFFFAOYSA-M
SMILES: [Na+].OCS([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results