accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HOO

CAS RN 149-57-5

IUPAC Name: 2-ethylhexanoic acid
Synonyms: 2-ethylhexanoic acid tridecyl ethylhexanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: OBETXYAYXDNJHR-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (7)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)