accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHOHHO

CAS RN 149-91-7

IUPAC Name: 3,4,5-trihydroxybenzoic acid
Synonyms: galop gallic acid tech. tech. gallic acid gallic acid, tech. kyselina gallova acid, gallic gallic acid polymer
Molecular Weight (g/mol): 170.12
Molecular Formula: C7H6O5
InChi Key: LNTHITQWFMADLM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C(O)C(O)=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)