accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 149022-15-1

IUPAC Name: 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
Synonyms: 2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
Molecular Weight (g/mol): 215.7
Molecular Formula: C8H10ClN3S
InChi Key: OEZPVSPULCMUQB-UHFFFAOYSA-N
SMILES: Cl.CN1C(SC2=CC=CC=C12)=NN
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results