accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClNNPt

CAS RN 14913-33-8

IUPAC Name: platinum(2+) diamine dichloride
Synonyms: cisplatin
Molecular Weight (g/mol): 300.05
Molecular Formula: Cl2H6N2Pt
InChi Key: LXZZYRPGZAFOLE-UHFFFAOYSA-L
SMILES: N.N.[Cl-].[Cl-].[Pt++]
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)