accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HCN

CAS RN 14918-21-9

IUPAC Name: hex-5-ynenitrile
Synonyms: hex-5-ynenitrile
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: JZYKFLLRVPPISG-UHFFFAOYSA-N
SMILES: C#CCCCC#N
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results