accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1492-30-4

IUPAC Name: 4-nitrophenyl hexadecanoate
Synonyms: 4-nitrophenyl hexadecanoate
Molecular Weight (g/mol): 377.53
Molecular Formula: C22H35NO4
InChi Key: LVZSQWIWCANHPF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results