accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSOOFFF

CAS RN 1493-13-6

IUPAC Name: barium(2+) ditrifluoromethanesulfonate
Synonyms: trifluoromethanesulfonic acid
Molecular Weight (g/mol): 435.45
Molecular Formula: C2BaF6O6S2
InChi Key: DXJURUJRANOYMX-UHFFFAOYSA-L
SMILES: [Ba++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (5)
  • (5)
  • (5)