accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH

CAS RN 14938-35-3

IUPAC Name: 4-pentylphenol
Synonyms: 4-n-pentylphenol amyl p-hydroxybenzene
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: ZNPSUQQXTRRSBM-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)