accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14947-53-6

IUPAC Name: 2-[2-(1,3-dithian-2-yl)ethyl]-1,3-dithiane
Synonyms: 2-[2-(1,3-dithian-2-yl)ethyl]-1,3-dithiane
Molecular Weight (g/mol): 266.49
Molecular Formula: C10H18S4
InChi Key: RVIPSMFHRZBJRI-UHFFFAOYSA-N
SMILES: C(CC1SCCCS1)C1SCCCS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results